Learn More
Waters Corp CORTECS T3 Column, 120Å, 2.7 μm, 4.6 mm X 30 mm
Catalog No. 502391714 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS™ Solid-Core Particle Columns enhance UHPLC separation by maximizing efficiency, increasing resolution, and improving throughput. These T3 columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. Their particle morphology provides high efficiency, lower backpressure, ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Reduces operational backpressure by 25% compared to fully porous packing materials of similar particle sizes.
  • Achieves higher throughput, up to 5-fold faster than conventional 5 μm HPLC columns.
  • Increases efficiency and resolution, especially beneficial for complex samples.
  • Maximizes asset utilization across existing HPLC, UHPLC, or UPLC systems.
Catalog No. 50-239-1714 Supplier Waters Corp Supplier No. 186008491
CORTECS™ Solid-Core Particle Columns enhance UHPLC separation by maximizing efficiency, increasing resolution, and improving throughput. These T3 columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. Their particle morphology provides high efficiency, lower backpressure, ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Reduces operational backpressure by 25% compared to fully porous packing materials of similar particle sizes.
  • Achieves higher throughput, up to 5-fold faster than conventional 5 μm HPLC columns.
  • Increases efficiency and resolution, especially beneficial for complex samples.
  • Maximizes asset utilization across existing HPLC, UHPLC, or UPLC systems.