Learn More
Waters Corp CORTECS T3 Column, 120Å, 2.7 μm, 2.1 mm ID x 150 mm, 3pk
Catalog No. 502391316 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product offers a CORTECS T3 Column, provided in a pack of three.
  • Pore size of 120 Å.
  • Utilizes 2.7 μm particle size.
  • Internal diameter (ID) of 2.1 mm and a length of 150 mm.
  • Employs the CORTECS T3 phase.
  • Classified as an AQ phase type.
  • Designed as a column.
  • Conforms to USP L1 standards.
Catalog No. 50-239-1316 Supplier Waters Corp Supplier No. 176003880
This product offers a CORTECS T3 Column, provided in a pack of three.
  • Pore size of 120 Å.
  • Utilizes 2.7 μm particle size.
  • Internal diameter (ID) of 2.1 mm and a length of 150 mm.
  • Employs the CORTECS T3 phase.
  • Classified as an AQ phase type.
  • Designed as a column.
  • Conforms to USP L1 standards.