Learn More
Waters Corp Waters Column, T3, 120Å, 2.7μm, 3 mm x 100 mm
Catalog No. 502224206 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size. CORTECS T3 provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases for polar and non-polar analytes.
  • Solid Core particle technology provides high efficiency and lower backpressure.
  • Ensures ultra-low column MS bleed and excellent peak shape.
  • Offers long lifetimes for reliable, reproducible results.
  • Increases throughput, efficiency, and resolution, decreasing analysis costs.
Catalog No. 50-222-4206 Supplier Waters Corp Supplier No. 186008489
CORTECS T3 columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure compared to fully porous particles of similar size. CORTECS T3 provides ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases for polar and non-polar analytes.
  • Solid Core particle technology provides high efficiency and lower backpressure.
  • Ensures ultra-low column MS bleed and excellent peak shape.
  • Offers long lifetimes for reliable, reproducible results.
  • Increases throughput, efficiency, and resolution, decreasing analysis costs.