Learn More
Waters Corp Waters CORTECS T3 Column, 120 Angstrom, 2.7 Micrometer
Supplier:  Waters Corp 186008484
Encompass_Preferred
CORTECS T3 columns feature Solid Core particle technology, offering high efficiency, lower backpressure, and compatibility with 100% aqueous mobile phases. They provide excellent retention for polar and non-polar analytes, increasing throughput across HPLC, UHPLC, or UPLC systems.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Offers high efficiency and lower backpressure compared to fully porous particles of similar size.
  • Delivers ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Designed with Solid Core particle technology.
  • Connects with any HPLC, UHPLC, or UPLC system.
  • Enables higher efficiency and increased throughput with enhanced resolution.
  • Allows for faster sample analysis, reducing laboratory operational costs.
  • Manufactured under strict controls for reproducible results.
Catalog No. 50-222-4205