Learn More
Waters Corp CORTECS UPLC Method Validation Kit, T3, 2.7 μm, 2.1 × 75 mm
Catalog No. 502390746 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS UPLC Columns Method Validation Kits are designed to help evaluate the quality, reliability, and consistency of an analytical method. These kits ensure the reproducibility of a chromatographic column's performance, which is crucial for the long-term reliability and robustness of an analytical method. Controlled particle and column manufacturing processes guarantee batch-to-batch and column-to-column reproducibility.
  • Kit includes three batches of chromatographic media.
  • Each batch is derived from different base particles.
  • Designed to help evaluate the quality, reliability, and consistency of an analytical method.
  • Ensures reproducibility of column performance.
  • Manufacturing processes guarantee batch-to-batch and column-to-column reproducibility.
Catalog No. 50-239-0746 Supplier Waters Corp Supplier No. 186008516
CORTECS UPLC Columns Method Validation Kits are designed to help evaluate the quality, reliability, and consistency of an analytical method. These kits ensure the reproducibility of a chromatographic column's performance, which is crucial for the long-term reliability and robustness of an analytical method. Controlled particle and column manufacturing processes guarantee batch-to-batch and column-to-column reproducibility.
  • Kit includes three batches of chromatographic media.
  • Each batch is derived from different base particles.
  • Designed to help evaluate the quality, reliability, and consistency of an analytical method.
  • Ensures reproducibility of column performance.
  • Manufacturing processes guarantee batch-to-batch and column-to-column reproducibility.