Learn More
Waters Corp CORTECS T3 Method Validation Kit, 120Å, 2.7 μm, 2.1 x 30 mm
Catalog No. 502390954 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Method validation kits provide columns of three different batches. These columns are compatible with 100% aqueous mobile phases and offer excellent retention for polar and non-polar analytes. The particle morphology ensures high efficiency and lower backpressure compared to fully porous particles of similar size. They also provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Particle morphology offers high efficiency and lower backpressure.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.
  • Kits contain columns of three different batches for method validation.
Catalog No. 50-239-0954 Supplier Waters Corp Supplier No. 186008509
Method validation kits provide columns of three different batches. These columns are compatible with 100% aqueous mobile phases and offer excellent retention for polar and non-polar analytes. The particle morphology ensures high efficiency and lower backpressure compared to fully porous particles of similar size. They also provide ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Particle morphology offers high efficiency and lower backpressure.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.
  • Kits contain columns of three different batches for method validation.