Learn More
Waters Corp CORTECS T3 Method Validation Kit, 120Å, 1.6 μm, 3 mm X 150 mm
Catalog No. 502391503 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit provides columns from three different batches, designed for UPLC/UHPLC systems. The columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes. Featuring a unique particle morphology, they deliver high efficiency and lower backpressure compared to fully porous particles of similar size.
  • Compatible with 100% aqueous mobile phases.
  • Excellent retention for polar and non-polar analytes.
  • High efficiency and lower backpressure.
  • Ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • C18 chemistry with T3 bonding.
  • Optimized for UPLC/UHPLC systems.
  • 1.6 μm particle size and 120 Å pore size.
  • 3 mm inner diameter and 150 mm length.
  • Supplied as a 3-pack.
Catalog No. 50-239-1503 Supplier Waters Corp Supplier No. 186008542
This method validation kit provides columns from three different batches, designed for UPLC/UHPLC systems. The columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes. Featuring a unique particle morphology, they deliver high efficiency and lower backpressure compared to fully porous particles of similar size.
  • Compatible with 100% aqueous mobile phases.
  • Excellent retention for polar and non-polar analytes.
  • High efficiency and lower backpressure.
  • Ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • C18 chemistry with T3 bonding.
  • Optimized for UPLC/UHPLC systems.
  • 1.6 μm particle size and 120 Å pore size.
  • 3 mm inner diameter and 150 mm length.
  • Supplied as a 3-pack.