Learn More
Waters Corp Method Validation Kit, 1.6 μm, 120 Å, 3 x 30 mm, 3/pk
Catalog No. 502391927 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit provides columns of three different batches, designed for high-performance liquid chromatography. These columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. They deliver high efficiency and lower backpressure, ensuring ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Three distinct column batches for method validation
  • Compatible with 100% aqueous mobile phases
  • Excellent retention for polar and non-polar analytes
  • High efficiency and lower backpressure
  • Ultra-low column MS bleed
  • Ensures long lifetimes and excellent peak shape
  • C18 chemistry with T3 bonding technology
  • 1.6 μm particle size, 120 Å pore size
  • 3 mm inner diameter, 30 mm length
  • UPLC and UHPLC system compatible
  • 3 units per package
Catalog No. 50-239-1927 Supplier Waters Corp Supplier No. 186008538
This method validation kit provides columns of three different batches, designed for high-performance liquid chromatography. These columns are compatible with 100% aqueous mobile phases, offering excellent retention for polar and non-polar analytes. They deliver high efficiency and lower backpressure, ensuring ultra-low column MS bleed, long lifetimes, and excellent peak shape.
  • Three distinct column batches for method validation
  • Compatible with 100% aqueous mobile phases
  • Excellent retention for polar and non-polar analytes
  • High efficiency and lower backpressure
  • Ultra-low column MS bleed
  • Ensures long lifetimes and excellent peak shape
  • C18 chemistry with T3 bonding technology
  • 1.6 μm particle size, 120 Å pore size
  • 3 mm inner diameter, 30 mm length
  • UPLC and UHPLC system compatible
  • 3 units per package