Learn More
Waters Corp HPLC Column, CORTECS T3 Method Validation Kit, 120 Å, 2.7 μm, 30 mm x 3 mm
Catalog No. 502392267 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The HPLC Column CORTECS T3 Method Validation Kit is a high-quality liquid chromatography product. It is designed to assist in method validation processes, providing precise separation capabilities with its specific particle and pore sizes, along with optimized dimensions for reliable analytical results.
  • High-quality liquid chromatography product.
  • Suitable for method validation.
  • Features 2.7 μm particle size.
  • Offers 120 Å pore size.
  • Provided in a 30 mm x 3 mm format.
  • Available in a 3-pack.
Catalog No. 50-239-2267 Supplier Waters Corp Supplier No. 186008519
The HPLC Column CORTECS T3 Method Validation Kit is a high-quality liquid chromatography product. It is designed to assist in method validation processes, providing precise separation capabilities with its specific particle and pore sizes, along with optimized dimensions for reliable analytical results.
  • High-quality liquid chromatography product.
  • Suitable for method validation.
  • Features 2.7 μm particle size.
  • Offers 120 Å pore size.
  • Provided in a 30 mm x 3 mm format.
  • Available in a 3-pack.