Learn More
Waters Corp CORTECS T3 Method Validation Kit, 120Å, 2.7 μm, 4.6 mm X 75 mm
Catalog No. 502390830 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 method validation kits provide columns of three different batches. These columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure when compared to fully porous particles of similar particle size.
  • Provides columns of three different batches.
  • Compatible with 100% aqueous mobile phases.
  • Offers excellent retention for polar and non-polar analytes.
  • Features particle morphology for high efficiency and lower backpressure.
  • Delivers ultra-low column MS bleed.
  • Ensures long lifetimes.
  • Provides excellent peak shape.
Catalog No. 50-239-0830 Supplier Waters Corp Supplier No. 186008526
CORTECS T3 method validation kits provide columns of three different batches. These columns are compatible with 100% aqueous mobile phases and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficiency and lower backpressure when compared to fully porous particles of similar particle size.
  • Provides columns of three different batches.
  • Compatible with 100% aqueous mobile phases.
  • Offers excellent retention for polar and non-polar analytes.
  • Features particle morphology for high efficiency and lower backpressure.
  • Delivers ultra-low column MS bleed.
  • Ensures long lifetimes.
  • Provides excellent peak shape.