Learn More
Waters Corp CORTECS T3 Method Validation Kit, 120Å, 2.7 μm, 3 mm × 100 mm, 3 pk
Catalog No. 502390938 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 method validation kits contain columns of three batches, compatible with 100% aqueous mobile phases. They offer excellent retention for polar and non-polar analytes. Particle morphology ensures high efficiency, lower backpressure, ultra-low column MS bleed, long lifetimes, and excellent peak shape. Digital COAs and eCords are online at www.waters.com/coa.
  • Includes three column batches for method validation.
  • Compatible with 100% aqueous mobile phases.
  • Excellent retention for polar and non-polar analytes.
  • High efficiency and lower backpressure (compared to fully porous particles).
  • Ultra-low column MS bleed.
  • Long lifetimes and excellent peak shape.
  • Digital COAs/eCords online (www.waters.com/coa) for less waste.
Catalog No. 50-239-0938 Supplier Waters Corp Supplier No. 186008522
CORTECS T3 method validation kits contain columns of three batches, compatible with 100% aqueous mobile phases. They offer excellent retention for polar and non-polar analytes. Particle morphology ensures high efficiency, lower backpressure, ultra-low column MS bleed, long lifetimes, and excellent peak shape. Digital COAs and eCords are online at www.waters.com/coa.
  • Includes three column batches for method validation.
  • Compatible with 100% aqueous mobile phases.
  • Excellent retention for polar and non-polar analytes.
  • High efficiency and lower backpressure (compared to fully porous particles).
  • Ultra-low column MS bleed.
  • Long lifetimes and excellent peak shape.
  • Digital COAs/eCords online (www.waters.com/coa) for less waste.