Learn More
Waters Corp HPLC Column, CORTECS T3-MV Kit, C18, 2.7 μm, 4.6 × 50 mm, 3/pk
Catalog No. 502390823 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS UPLC Columns Method Validation Kit is designed to assess the reproducibility of a chromatographic column’s performance. It is crucial for the long-term reliability and robustness of an analytical method. These kits include three columns from three different batches of material to evaluate the quality, reliability, and consistency of your method.
  • Columns leverage highly controlled particle and column manufacturing processes to ensure batch-to-batch and column-to-column reproducibility.
  • Particle size of 2.7 μm.
  • Column dimensions of 4.6 × 50 mm.
  • Includes three columns per package.
  • Features C18 chemistry.
  • Pore size of 120 Å.
Catalog No. 50-239-0823 Supplier Waters Corp Supplier No. 186008525
This CORTECS UPLC Columns Method Validation Kit is designed to assess the reproducibility of a chromatographic column’s performance. It is crucial for the long-term reliability and robustness of an analytical method. These kits include three columns from three different batches of material to evaluate the quality, reliability, and consistency of your method.
  • Columns leverage highly controlled particle and column manufacturing processes to ensure batch-to-batch and column-to-column reproducibility.
  • Particle size of 2.7 μm.
  • Column dimensions of 4.6 × 50 mm.
  • Includes three columns per package.
  • Features C18 chemistry.
  • Pore size of 120 Å.