Learn More
Waters Corp CORTECS T3 Method Validation Kit, 120Å, 2.7 μm, 2.1 × 50 mm, 3/pk
Catalog No. 502390957 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 method validation kits are designed to assess the quality, reliability, and consistency of an analytical method by including three batches of chromatographic media. These columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes, combined with high efficiency and lower backpressure compared to fully porous particles.
  • Kit includes columns from three different batches.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Particle morphology offers high efficiency.
  • Lower backpressure compared to fully porous particles of similar size.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.
Catalog No. 50-239-0957 Supplier Waters Corp Supplier No. 186008510
CORTECS T3 method validation kits are designed to assess the quality, reliability, and consistency of an analytical method by including three batches of chromatographic media. These columns are compatible with 100% aqueous mobile phases and offer excellent retention for both polar and non-polar analytes, combined with high efficiency and lower backpressure compared to fully porous particles.
  • Kit includes columns from three different batches.
  • Compatible with 100% aqueous mobile phases.
  • Provides excellent retention for polar and non-polar analytes.
  • Particle morphology offers high efficiency.
  • Lower backpressure compared to fully porous particles of similar size.
  • Ultra-low column MS bleed.
  • Long lifetimes.
  • Excellent peak shape.