Learn More
Waters Corp CORTECS T3 Method Validation Kit, 120Å, 2.7 μm, 4.6 mm × 30 mm
Catalog No. 502390986 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS T3 method validation kits contain columns in three batches, compatible with 100% aqueous mobile phases. They offer excellent retention for polar and non-polar analytes. Their particle morphology provides high efficiency, lower backpressure, ultra-low MS bleed, long lifetimes, and excellent peak shape.
  • C18 chemistry.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range 2 to 8.
  • Maximum pressure 18000 psi (1240 Bar).
  • Endcapped.
  • T3 bonding technology.
  • Low silanol activity.
  • Spherical particle shape.
  • Parker-style endfitting type.
  • Method validation kit format.
  • UPLC and UHPLC system compatible.
  • Solid core particle technology.
  • L1 USP classification.
  • 4.7% carbon load.
  • 120Å pore size.
  • 4.6 mm inner diameter.
  • 30 mm length.
  • 3 units per package.
Catalog No. 50-239-0986 Supplier Waters Corp Supplier No. 186008524
CORTECS T3 method validation kits contain columns in three batches, compatible with 100% aqueous mobile phases. They offer excellent retention for polar and non-polar analytes. Their particle morphology provides high efficiency, lower backpressure, ultra-low MS bleed, long lifetimes, and excellent peak shape.
  • C18 chemistry.
  • Reversed phase separation mode.
  • Silica particle substrate.
  • pH range 2 to 8.
  • Maximum pressure 18000 psi (1240 Bar).
  • Endcapped.
  • T3 bonding technology.
  • Low silanol activity.
  • Spherical particle shape.
  • Parker-style endfitting type.
  • Method validation kit format.
  • UPLC and UHPLC system compatible.
  • Solid core particle technology.
  • L1 USP classification.
  • 4.7% carbon load.
  • 120Å pore size.
  • 4.6 mm inner diameter.
  • 30 mm length.
  • 3 units per package.