Learn More
Waters Corp Waters Corporation VanGuard Cartridge, 120Å, 2.7 μm, 3.9 mm × 5 mm
Catalog No. 502038609 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. They are optimized to protect analytical columns containing 2.7 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination.
  • Made with the same high-performance packing materials as analytical columns.
  • Tested chromatographically with acidic, basic, and neutral analytes.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Each pre-column is tested and approved for its specific use.
Catalog No. 50-203-8609 Supplier Waters Corp Supplier No. 186008507
These VanGuard Cartridges extend analytical column lifetime and performance by removing particulate contamination from the mobile phase stream. They are optimized to protect analytical columns containing 2.7 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination.
  • Made with the same high-performance packing materials as analytical columns.
  • Tested chromatographically with acidic, basic, and neutral analytes.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Each pre-column is tested and approved for its specific use.