Learn More
Waters Corp CORTECS T3 VanGuard Cartridge, 120Å, 2.7 μm, 2.1 mm × 5 mm
Catalog No. 502390939 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination. This 2.1 mm × 5 mm cartridge, with 2.7 μm sorbent particles, acts as a protective barrier against chemical contaminants and suspended solids, optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns.
  • Extends column lifetime and protects against deterioration.
  • Ensures compatibility and performance with analytical columns.
  • Protects against chemical contaminants and suspended solids.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Chromatographically tested for repeatable performance.
  • Approved for improving selectivity, sensitivity, and analysis speed.
  • Suitable for biological fluid purification or temperature control.
Catalog No. 50-239-0939 Supplier Waters Corp Supplier No. 186008506
VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination. This 2.1 mm × 5 mm cartridge, with 2.7 μm sorbent particles, acts as a protective barrier against chemical contaminants and suspended solids, optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns.
  • Extends column lifetime and protects against deterioration.
  • Ensures compatibility and performance with analytical columns.
  • Protects against chemical contaminants and suspended solids.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Chromatographically tested for repeatable performance.
  • Approved for improving selectivity, sensitivity, and analysis speed.
  • Suitable for biological fluid purification or temperature control.