Learn More
Waters Corp Waters VanGuard Pre-column, 120Å, 1.6 μm, 2.1 mm × 5 mm
Catalog No. 502038610 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
VanGuard Pre-Columns extend analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase stream, protecting UPLC T3 analytical columns with 1.6 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Optimized for UPLC T3 analytical columns with 1.6 μm sorbent particles.
  • Uses same packing material as analytical column for compatibility.
  • Removes chemical impurities and suspended solids.
  • Manufactured in a cGMP, ISO 9001 certified plant with high-performance materials.
  • Designed for increased capacity in biological fluid cleanup and temperature control.
  • Chromatographically tested batches ensure reproducible performance.
  • Each pre-column is certified for optimal performance, selectivity, sensitivity, and speed.
Catalog No. 50-203-8610 Supplier Waters Corp Supplier No. 186008508
VanGuard Pre-Columns extend analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase stream, protecting UPLC T3 analytical columns with 1.6 μm sorbent particles.
  • Extends analytical column lifetime and performance.
  • Optimized for UPLC T3 analytical columns with 1.6 μm sorbent particles.
  • Uses same packing material as analytical column for compatibility.
  • Removes chemical impurities and suspended solids.
  • Manufactured in a cGMP, ISO 9001 certified plant with high-performance materials.
  • Designed for increased capacity in biological fluid cleanup and temperature control.
  • Chromatographically tested batches ensure reproducible performance.
  • Each pre-column is certified for optimal performance, selectivity, sensitivity, and speed.