Learn More
Waters Corp CORTECS Column, C18, 1.6 μm, 2.1 mm × 50 mm
Supplier:  Waters Corp 176003147
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • High-efficiency columns based on a solid-core particle.
  • Offer balanced retention of acids, bases and neutrals at low and mid-range pH.
  • Leverage the power of the sub-2-μm to deliver outstanding results, whether resolving complex mixtures or maintaining resolution.
  • Guarantees higher column efficiency.
  • Solid-core particles improve performance by lowering the van Deemter equation terms (A, B, and C).
Catalog No. 50-153-4798