Learn More
Waters Corp Waters CORTECS T3 Column, 1.6 μm, 2.1 x 150 mm, 120 Å
Catalog No. 501384923 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This CORTECS T3 column is a reversed-phase column designed for UPLC and UHPLC systems, known for its C18 chemistry and T3 bonding technology. Utilizing solid core particle technology, it delivers high efficiency and reduced backpressure, making it ideal for the separation of diverse analytes.
  • Compatible with 100% aqueous mobile phases.
  • Offers excellent retention for both polar and non-polar analytes.
  • Delivers high efficiency and lower backpressure.
  • Features ultra-low column MS bleed, extended lifetimes, and excellent peak shape.
  • 120Å pore diameter for 100% aqueous mobile phase use without phase dewetting.
  • Chemically stable at low pH and fully LC-MS compatible.
  • Solid-core technology reduces backpressure and increases sensitivity.
  • T3 bonding ensures superior polar compound retention and aqueous compatibility.
Catalog No. 50-138-4923 Supplier Waters Corp Supplier No. 186008500
Add to Cart
Add to Cart
This CORTECS T3 column is a reversed-phase column designed for UPLC and UHPLC systems, known for its C18 chemistry and T3 bonding technology. Utilizing solid core particle technology, it delivers high efficiency and reduced backpressure, making it ideal for the separation of diverse analytes.
  • Compatible with 100% aqueous mobile phases.
  • Offers excellent retention for both polar and non-polar analytes.
  • Delivers high efficiency and lower backpressure.
  • Features ultra-low column MS bleed, extended lifetimes, and excellent peak shape.
  • 120Å pore diameter for 100% aqueous mobile phase use without phase dewetting.
  • Chemically stable at low pH and fully LC-MS compatible.
  • Solid-core technology reduces backpressure and increases sensitivity.
  • T3 bonding ensures superior polar compound retention and aqueous compatibility.