Learn More
Waters Corp CORTECS T3 Column, 120Å, 1.6 µm, 2.1 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186008497
Encompass_Preferred
CORTECS T3 columns are compatible with 100% aqueous mobile phases, and provide excellent retention for polar and non-polar analytes. The particle morphology offers high efficnecy and lower backpressure when compared to fully porouse particles of similar particle size. CORTECS T3 provides you with ultra-low column MS bleed, long lifetimes and excellent peak shape.
Catalog No. 50-138-4920