Learn More
Waters Corp Waters Corporation C18 Column, 2.7 μm, 120 Å, 4.6 mm X 150 mm
Supplier:  Waters Corp 186008495
Encompass_Preferred
These columns are designed for high-speed applications, offering excellent retention for both polar and non-polar analytes even with 100% aqueous mobile phases. Their advanced particle morphology ensures high efficiency and reduced backpressure.
  • Ultra-low column mass spectrometry bleed.
  • Long lifetimes and excellent peak shape.
  • Reproducible and superior retention for polar and non-polar analytes.
  • Enhanced polar interactions and retention through improved access to silanol groups.
  • Improved resistance to dewetting with low or no organic solvent mobile phases.
  • Optimized for high-performance liquid chromatography and ultra-performance liquid chromatography instruments.
  • Separates a wide range of compound classes.
  • The 2.7 μm particle size delivers sharper, narrower peaks and faster flow rates at lower backpressure.
Catalog No. 50-138-4918