Learn More
Waters Corp Waters CORTECS Solid Core C8 Column, 1.6 μm, 2.1 mm × 150 mm, 90 Å
Catalog No. 501384878 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
CORTECS C8 columns are designed for general purpose method development under low pH conditions. The C8 ligand is less hydrophobic and less retentive than C18 phases. Paired with low-dispersive UPLC equipment, 1.6 μm solid-core particle technology ensures superior column efficiency.
  • Designed for method development under low pH conditions.
  • C8 ligand offers reduced retention versus C18 phases.
  • Superior efficiency with 1.6 μm solid-core particle technology.
  • Generates sharper, narrower peaks.
  • Enhances resolution, speed, and sensitivity.
  • Solid-core particles with tri-functionally bound C8 Ligand.
  • Shorter alkyl chain reduces hydrophobicity and retention.
  • Reduces analysis time and solvent expenses for hydrophobic analytes.
  • Manufactured under stringent guidelines with rigorous quality assurance.
Catalog No. 50-138-4878 Supplier Waters Corp Supplier No. 186008402
CORTECS C8 columns are designed for general purpose method development under low pH conditions. The C8 ligand is less hydrophobic and less retentive than C18 phases. Paired with low-dispersive UPLC equipment, 1.6 μm solid-core particle technology ensures superior column efficiency.
  • Designed for method development under low pH conditions.
  • C8 ligand offers reduced retention versus C18 phases.
  • Superior efficiency with 1.6 μm solid-core particle technology.
  • Generates sharper, narrower peaks.
  • Enhances resolution, speed, and sensitivity.
  • Solid-core particles with tri-functionally bound C8 Ligand.
  • Shorter alkyl chain reduces hydrophobicity and retention.
  • Reduces analysis time and solvent expenses for hydrophobic analytes.
  • Manufactured under stringent guidelines with rigorous quality assurance.