Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters CORTECS C8 Column, 90Å, 1.6 μm, 2.1 × 50 mm

Catalog No. 501384875 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This CORTECS C8 Column provides increased resolution and enhanced efficiency for challenging liquid chromatography samples. Designed for general-purpose method development under low pH, it delivers reproducible results across multiple assays.

  • C8 chemistry for reversed phase separation.
  • Silica substrate; spherical, solid core particles.
  • PH range: 2 to 8.
  • Temperature limit: 45°C.
  • Maximum pressure: 18000 psi (1240 Bar).
  • Low silanol activity, endcapped.
  • Particle size: 1.6 μm.
  • Pore size: 90Å.
  • Parker-style endfitting.
  • Surface area: 100.
  • For UPLC and UHPLC systems.
  • USP classification: L7.
  • Dimensions: 2.1 mm inner diameter, 50 mm length.
  • Carbon load: 4.5%.
  • Equipped with eCord.

Catalog No. 50-138-4875 Supplier Waters Corp Supplier No. 186008399
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This CORTECS C8 Column provides increased resolution and enhanced efficiency for challenging liquid chromatography samples. Designed for general-purpose method development under low pH, it delivers reproducible results across multiple assays.

  • C8 chemistry for reversed phase separation.
  • Silica substrate; spherical, solid core particles.
  • PH range: 2 to 8.
  • Temperature limit: 45°C.
  • Maximum pressure: 18000 psi (1240 Bar).
  • Low silanol activity, endcapped.
  • Particle size: 1.6 μm.
  • Pore size: 90Å.
  • Parker-style endfitting.
  • Surface area: 100.
  • For UPLC and UHPLC systems.
  • USP classification: L7.
  • Dimensions: 2.1 mm inner diameter, 50 mm length.
  • Carbon load: 4.5%.
  • Equipped with eCord.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.