Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters CORTECS C8 Column, 90Å, 2.7 μm

Catalog No. 501384848 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

CORTECS C8 columns are designed for general-purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and thus less retentive compared to C18 phases, making it preferred for certain assays. These columns feature solid-core particles with a tri-functionally bonded C8 Ligand.

  • Offers shorter retention times for hydrophobic and extremely hydrophobic analytes.
  • Provides high column efficiencies with low-dispersive UPLC instrumentation.
  • Features 2.7 μm particles for enhanced resolution, speed, and sensitivity due to sharp, narrower peaks.
  • Withstands pressure up to 18,000 psi (1240 Bar).
  • Maximum operating temperature of 45 °C.
  • pH range of 2 to 8.
  • Endcapped for optimal performance.
  • Low silanol activity.
  • Spherical particle shape.

Catalog No. 50-138-4848 Supplier Waters Corp Supplier No. 186008372
May include imposed supplier surcharges.
Add to Cart
Add to Cart

CORTECS C8 columns are designed for general-purpose method development under low pH test conditions. The C8 ligand is less hydrophobic and thus less retentive compared to C18 phases, making it preferred for certain assays. These columns feature solid-core particles with a tri-functionally bonded C8 Ligand.

  • Offers shorter retention times for hydrophobic and extremely hydrophobic analytes.
  • Provides high column efficiencies with low-dispersive UPLC instrumentation.
  • Features 2.7 μm particles for enhanced resolution, speed, and sensitivity due to sharp, narrower peaks.
  • Withstands pressure up to 18,000 psi (1240 Bar).
  • Maximum operating temperature of 45 °C.
  • pH range of 2 to 8.
  • Endcapped for optimal performance.
  • Low silanol activity.
  • Spherical particle shape.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.