Learn More
Waters Corp Waters CORTECS T3 Column, 2.7 μm, 120 Å, 4.6 mm ID, 100 mm L
Catalog No. NC1835939 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These CORTECS T3 columns are designed for high-speed applications, offering compatibility with 100% aqueous mobile phases and excellent retention for both polar and non-polar analytes. Their unique particle morphology ensures high efficiency and reduced backpressure compared to fully porous particles of similar size, facilitating faster and more precise analytical results.
  • Compatible with 100% aqueous mobile phases
  • Provides excellent retention for both polar and non-polar analytes
  • Offers high efficiency and lower backpressure compared to fully porous particles
  • Features ultra-low column MS bleed
  • Ensures long lifetimes and excellent peak shape
  • Boosts efficiency and increases throughput with higher flow rates
  • Improves resolution for complex sample analysis
  • Manufactured in an ISO 9001-certified facility
Catalog No. NC1835939 Supplier Waters Corp Supplier No. 186008494
These CORTECS T3 columns are designed for high-speed applications, offering compatibility with 100% aqueous mobile phases and excellent retention for both polar and non-polar analytes. Their unique particle morphology ensures high efficiency and reduced backpressure compared to fully porous particles of similar size, facilitating faster and more precise analytical results.
  • Compatible with 100% aqueous mobile phases
  • Provides excellent retention for both polar and non-polar analytes
  • Offers high efficiency and lower backpressure compared to fully porous particles
  • Features ultra-low column MS bleed
  • Ensures long lifetimes and excellent peak shape
  • Boosts efficiency and increases throughput with higher flow rates
  • Improves resolution for complex sample analysis
  • Manufactured in an ISO 9001-certified facility