Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Armstrong Ceilings Ceiling Tile, Mineral Fiber, 24 x 24 in, 5/8 in Thick, Beveled Tegular

Catalog No. 502836900
Encompass

This ceiling tile provides an economical solution for commercial spaces, featuring a medium-textured, mineral fiber construction. Designed for suspended grid installations, it offers standard acoustical absorption and resistance to mold and mildew. Its non-directional visual and beveled tegular edge contribute to efficient installation and a clean aesthetic.

  • Medium-textured, economical solution
  • Standard acoustical absorption (NRC 0.55, CAC 35)
  • Beveled tegular edge detail minimizes the look of grid
  • Non-directional visual reduces scrap and installation time
  • Constructed from mineral fiber
  • Installs in suspended grid systems
  • Resists growth of mold and mildew on the panel surface
  • Class A fire performance
  • 80% light reflectance
  • Manufactured in the U.S.A.
  • Contains up to 41% recycled content
  • Recyclable through a dedicated program

Catalog No. 50-283-6900 Supplier MSC Supplier No. 49023344
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ceiling tile provides an economical solution for commercial spaces, featuring a medium-textured, mineral fiber construction. Designed for suspended grid installations, it offers standard acoustical absorption and resistance to mold and mildew. Its non-directional visual and beveled tegular edge contribute to efficient installation and a clean aesthetic.

  • Medium-textured, economical solution
  • Standard acoustical absorption (NRC 0.55, CAC 35)
  • Beveled tegular edge detail minimizes the look of grid
  • Non-directional visual reduces scrap and installation time
  • Constructed from mineral fiber
  • Installs in suspended grid systems
  • Resists growth of mold and mildew on the panel surface
  • Class A fire performance
  • 80% light reflectance
  • Manufactured in the U.S.A.
  • Contains up to 41% recycled content
  • Recyclable through a dedicated program

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.