Learn More
Cayman Chemical Cortexolone 17α-propionate, Purity: ≥98%, CAS: 19608-29-8, 10 mg
SDP
Catalog No. 502940486
Encompass_Preferred
A peripherally selective androgen receptor antagonist; binds to AR and inhibits AR-regulated transcription in a reporter cell line in vitro; inhibits androgen-regulated lipid synthesis in primary human sebocytes; inhibits inflammatory cytokine production from primary human sebocytes and scalp dermal papilla cells; inhibits testosterone propionate-induced flank organ enlargement in Syrian golden hamsters by 84% when administered topically at 400 μg/animal
Catalog No. 50-294-0486 Supplier Cayman Chemical Supplier No. 2839210 MG
A peripherally selective androgen receptor antagonist; binds to AR and inhibits AR-regulated transcription in a reporter cell line in vitro; inhibits androgen-regulated lipid synthesis in primary human sebocytes; inhibits inflammatory cytokine production from primary human sebocytes and scalp dermal papilla cells; inhibits testosterone propionate-induced flank organ enlargement in Syrian golden hamsters by 84% when administered topically at 400 μg/animal