Learn More
Pyramex™ Cortez™ Safety Eyewear
Catalog No. 19155702 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor, Mirror
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-702 Clear
19-155-703 Gray
19-155-704 Indoor/Outdoor, Mirror
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-702 Supplier Pyramex™ Supplier No. SB3610ST
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
A well-balanced pair of safety glasses suitable for a variety of tasks
  • Durable co-injected temples
  • Ribbed cushioned nosepiece
  • Specifications
    Lens Coating Anti-fog
    Lens Type H2X
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Clear
    Frame Color Black
    Lens Material Polycarbonate