Learn More
Brainxell Inc Cortical Astrocytes (Human iPSC-Derived, Male Line, 2M)
SDP
Supplier:  Brainxell Inc BX0600302M
Encompass
Cortical astrocytes derived from iPSC line WC-30, male. Contents: 1 vial of ≥2x10^6 viable cryopreserved cells. Marker Expression and Morphology: Cortical Astrocytes (BX-0600) express key astrocyte markers GFAP (green) at >70%. Cortical Astrocytes promote neurite outgrowth and exhibit mature process-bearing morphologies (shown with a DAPI stain). Function: Cortical Astrocytes uptake glutamate. Cortical Astrocytes exhibit calcium waves. Related diseases: Autism, Epilepsy, Alzheimer's & Huntington's Disease.
Catalog No. NC1991255