Learn More
Brainxell Inc Cortical Glutamatergic Neurons, 1M Viable Cells/Vial - Standard (BX-0300e)
SDP
Supplier:  Brainxell Inc BX0300301M/DUP
Encompass
Origin: Human iPSC lineContents: 1 vial of cells and corresponding BrainXell supplementsCryopreserved
Catalog No. NC3001389