Learn More
Med Vet International Cortical Nut, 4.5mm, J0375C, Each
Catalog No. 5000581530
Encompass_Preferred
Occasionally tapped threads strip and a larger screw is inappropriate. Lag screws are particularly vulnerable, as only one cortex is available for tapping. A cortical nut manufactured from implant grade nylon can give the screw purchase to allow normal tightening. The thread form is designed to grip the screw to minimize backing out. Sterile.
Catalog No. 50-005-81530 Supplier Med Vet International Supplier No. JOR375C
Occasionally tapped threads strip and a larger screw is inappropriate. Lag screws are particularly vulnerable, as only one cortex is available for tapping. A cortical nut manufactured from implant grade nylon can give the screw purchase to allow normal tightening. The thread form is designed to grip the screw to minimize backing out. Sterile.