Learn More
Med Vet International Cortical Screw, J0416D, 2mm x 12mm, 6 Count
Catalog No. 5000581604
Encompass_Preferred
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.
Catalog No. 50-005-81604 Supplier Med Vet International Supplier No. JOR416D
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.