Learn More
Med Vet International Cortical Screw, J0419J, 2.7mm x 24mm, 6 Count
Catalog No. 5000582687
Encompass_Preferred
2.7mm Cortical Screw. Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site.
Catalog No. 50-005-82687 Supplier Med Vet International Supplier No. JOR419J
2.7mm Cortical Screw. Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site.