Learn More
Med Vet International Cortical Screws, J0419C, 2.7mm x 10mm, 6 Count
Catalog No. 5000581627
Encompass_Preferred
2.7mm Cortical Screw. Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site.
Catalog No. 50-005-81627 Supplier Med Vet International Supplier No. JOR419C
2.7mm Cortical Screw. Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site.