Learn More
Med Vet International Cortical Screws, J0419N, 2.7mm x 32mm, 6 Count
Catalog No. 5000581669
Encompass_Preferred
2.7mm Cortical Screw. Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site.
Catalog No. 50-005-81669 Supplier Med Vet International Supplier No. JOR419N
2.7mm Cortical Screw. Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site.