Learn More
Med Vet International Cortical Screws, J0421D, 1.25 Pitch, 3.5mm x 16mm, 6 Count
Catalog No. 5000582894
Encompass_Preferred
1.25 Pitch. Din 58812. Use with 3.5mm compression platesto obtain fracture-site compression. 6/pk. Self-Tapping: 10mm through 40 mm.
Catalog No. 50-005-82894 Supplier Med Vet International Supplier No. JOR421D
1.25 Pitch. Din 58812. Use with 3.5mm compression platesto obtain fracture-site compression. 6/pk. Self-Tapping: 10mm through 40 mm.