Learn More
Med Vet International Cortical Screws, J0421R, 1.25 Pitch, 3.5mm x 55mm, 6 Count
Catalog No. 5000581941
Encompass_Preferred
1.25 Pitch. Din 58812. Use with 3.5mm compression platesto obtain fracture-site compression. 6/pk. Self-Tapping: 10mm through 40 mm.
Catalog No. 50-005-81941 Supplier Med Vet International Supplier No. JOR421R
1.25 Pitch. Din 58812. Use with 3.5mm compression platesto obtain fracture-site compression. 6/pk. Self-Tapping: 10mm through 40 mm.