Learn More
Med Vet International Cortical Self-Tapping Screw, J0416AS, 2mm x 6mm, Each
Catalog No. 5000581685
Encompass_Preferred
2mm Cortical Screw. Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.
Catalog No. 50-005-81685 Supplier Med Vet International Supplier No. JOR416AS
2mm Cortical Screw. Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.