Learn More
Med Vet International Cortical Self-Tapping Screw, J0416CS, 2.0mm, 10mm, Each
Catalog No. 5000582093
Encompass_Preferred
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.
Catalog No. 50-005-82093 Supplier Med Vet International Supplier No. JOR416CS
Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.