Learn More
Med Vet International Cortical Self-Tapping Screw, J0416ES, 2mm x 14mm, Each
Catalog No. 5000582867
Encompass_Preferred
2mm Cortical Screw. Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.
Catalog No. 50-005-82867 Supplier Med Vet International Supplier No. JOR416ES
2mm Cortical Screw. Available in 6mm-24mm lengths, the spherical head of the screw produces compression when used in conjunction with 2mm compression plate.