Learn More
Med Vet International Cortical Self-Tapping Screw, J0419IS, 2.7mm x 22mm, Each
Catalog No. 5000582889
Encompass_Preferred
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-tapping threads. 22mm L. Stainless Steel.
Catalog No. 50-005-82889 Supplier Med Vet International Supplier No. JOR419IS
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-tapping threads. 22mm L. Stainless Steel.