Learn More
Med Vet International Cortical Self-Tapping Screw, J0419OS, 2.7mm x 34mm, Each
Catalog No. 5000581711
Encompass_Preferred
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-tapping threads. 34mm L. Stainless Steel.
Catalog No. 50-005-81711 Supplier Med Vet International Supplier No. JOR419OS
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-tapping threads. 34mm L. Stainless Steel.