Learn More
Med Vet International Cortical Self-Tapping Screw, J0419RS, 2.7mm x 40mm, Each
Catalog No. 5000582924
Encompass_Preferred
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-tapping threads. 40mm L. Stainless Steel.
Catalog No. 50-005-82924 Supplier Med Vet International Supplier No. JOR419RS
Use with 2.7mm compression and 2.7mm reconstruction plates to obtain compression at a fracture site. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-tapping threads. 40mm L. Stainless Steel.