Learn More
Med Vet International Cortical Self-Tapping Screw, J0421CS, 1.25 Pitch, 3.5mm x 14mm, Each
Catalog No. 5000582726
Encompass_Preferred
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-Tapping. 14mm L.
Catalog No. 50-005-82726 Supplier Med Vet International Supplier No. JOR421CS
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Screw. Self-Tapping. 14mm L.