Learn More
Med Vet International Cortical Self-Tapping Screw, J0421IS, 1.25 Pitch, 3.5mm x 26mm, Each
Catalog No. 5000582715
Encompass_Preferred
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO. Screw. Self-Tapping. 26mm L. Stainless Steel.
Catalog No. 50-005-82715 Supplier Med Vet International Supplier No. JOR421IS
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO. Screw. Self-Tapping. 26mm L. Stainless Steel.