Learn More
Med Vet International Cortical Self-Tapping Screw, J0421NS, 1.25 Pitch, 3.5mm x 36mm, Each
Catalog No. 5000582729
Encompass_Preferred
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO. Screw. Self-Tapping. 36mm L. Stainless Steel.
Catalog No. 50-005-82729 Supplier Med Vet International Supplier No. JOR421NS
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping threads sold in single packs.Self-Tapping screws save time and work extremely well in most circumstances especally in the proximal tibia for TPLO and TTO. Screw. Self-Tapping. 36mm L. Stainless Steel.