Learn More
Med Vet International Cortical Self-Tapping Screw, J0421PS, 1.25 Pitch, 3.5mm x 45mm, Each
Catalog No. 5000582718
Encompass_Preferred
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping screws save time and work extremely well in most cirbumstances especially in the proximal tibia fro TPLO and TTO. Available in 2.0, 2.4, 2.7 and 3.5
Catalog No. 50-005-82718 Supplier Med Vet International Supplier No. JOR421PS
1.25 Pitch. Din 58812. Use with 3.5mm compression plates to obtain fracture-site compression. Self-tapping screws save time and work extremely well in most cirbumstances especially in the proximal tibia fro TPLO and TTO. Available in 2.0, 2.4, 2.7 and 3.5