Learn More
Med Vet International Cortical Tap, J0409A, 2.0mm, Each
Catalog No. 5000582063
Encompass_Preferred
By carefully positioning cortical screws in the special holes in compression plates, compression can be exerted at the reduced fracture site as the screw is tightened. As the head of the screw strikes the edge of the plate’s hole, the screw (and the associated bone) are forced away from the edge of the hole and toward the fracture site. The object of compression is to provide for primary bone union. A special guide must be used to position the screw hole accurately in the plate. Holes for cortical screws must be pre-tapped prior to screw placement.
Catalog No. 50-005-82063 Supplier Med Vet International Supplier No. JOR409A
By carefully positioning cortical screws in the special holes in compression plates, compression can be exerted at the reduced fracture site as the screw is tightened. As the head of the screw strikes the edge of the plate’s hole, the screw (and the associated bone) are forced away from the edge of the hole and toward the fracture site. The object of compression is to provide for primary bone union. A special guide must be used to position the screw hole accurately in the plate. Holes for cortical screws must be pre-tapped prior to screw placement.