Learn More
Cayman Chemical Corticosterone, Purity: ≥98%, CAS: 50-22-6, 100 mg
SDP
Catalog No. 502972128
Encompass_Preferred
A steroid hormone produced in the cortex of the adrenal glands that binds to both glucocorticoid and mineralocorticoid receptors; also is the precursor to aldosterone synthesis
Catalog No. 50-297-2128 Supplier Cayman Chemical Supplier No. 16063100 MG
A steroid hormone produced in the cortex of the adrenal glands that binds to both glucocorticoid and mineralocorticoid receptors; also is the precursor to aldosterone synthesis