Learn More
Medchemexpress LLC Corticosterone | 50-22-6 | 99.8% | 100 MG
SDP
Catalog No. 5000411472
Encompass_Preferred
Corticosterone (17-Deoxycortisol) is an orally active glucocorticoid produced in the adrenal cortex. It regulates neuronal functions of the limbic system, including the hippocampus, prefrontal cortex, and amygdala. This compound interferes with the maturation of dendritic cells, exhibiting an immunosuppressive effect.
  • Regulates neuronal functions of the limbic system.
  • Increases Rab-mediated AMPAR membrane traffic.
  • Shows immunosuppressive effects by interfering with dendritic cell maturation.
  • Inhibits organic cation transporters (OCTs).
  • Enhances glutamatergic transmission and synaptic AMPAR currents.
Catalog No. 50-004-11472 Supplier Medchemexpress LLC Supplier No. HYB1618100MG
Corticosterone (17-Deoxycortisol) is an orally active glucocorticoid produced in the adrenal cortex. It regulates neuronal functions of the limbic system, including the hippocampus, prefrontal cortex, and amygdala. This compound interferes with the maturation of dendritic cells, exhibiting an immunosuppressive effect.
  • Regulates neuronal functions of the limbic system.
  • Increases Rab-mediated AMPAR membrane traffic.
  • Shows immunosuppressive effects by interfering with dendritic cell maturation.
  • Inhibits organic cation transporters (OCTs).
  • Enhances glutamatergic transmission and synaptic AMPAR currents.